• USA Home
  • 41765 - Dimethylzinc solution

EMAIL THIS PAGE TO A FRIEND
41765 Sigma-Aldrich

Dimethylzinc solution

purum, ~1 M in heptane

  • CAS Number 544-97-8

  • Linear Formula (CH3)2Zn

  • Molecular Weight 95.46

  •  Beilstein Registry Number 3587195

  •  MDL number MFCD00014854

  •  PubChem Substance ID 329756151

Purchase

Properties

InChI Key   AXAZMDOAUQTMOW-UHFFFAOYSA-N
grade   purum
concentration   ~1 M in heptane
density   0.72 g/mL at 20 °C

Description

Caution

may contain precipitate

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN3399 - class 4.3 - PG 2 - EHS - Organometallic substance, liquid, water-reactive, flammable, HI: all (not BR)
WGK Germany 
3
Flash Point(F) 
30.2 °F
Flash Point(C) 
-1 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

417246 Dimethylzinc solution, 1.0 M in heptane

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?