• USA Home
  • 41811 - 1,2-Dipalmitoyl-sn-glycero-3-phosphate calcium salt

EMAIL THIS PAGE TO A FRIEND
41811 Sigma-Aldrich

1,2-Dipalmitoyl-sn-glycero-3-phosphate calcium salt

≥98.0% (TLC)

Synonym: 3-sn-Phosphatidic acid, 1,2-dipalmitoyl calcium salt, L-β,γ-Dipalmitoyl-α-phosphatidic acid calcium salt

  • CAS Number 71640-91-0

  • Empirical Formula (Hill Notation) C35H67CaO8P

  • Molecular Weight 686.95

  •  MDL number MFCD00055713

Purchase

Properties

Related Categories Biochemicals and Reagents, Glycerophosphates, Glycerophospholipids, Lipids
InChI Key   JICVBQGRHKLMHG-MGDILKBHSA-L
assay   ≥98.0% (TLC)
storage temp.   −20°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?