• USA Home
  • 41964 - O-[2-(6-Oxocaproylamino)ethyl]-O′-methylpolyethylene glycol 5′000

EMAIL THIS PAGE TO A FRIEND
41964 Sigma-Aldrich

O-[2-(6-Oxocaproylamino)ethyl]-O′-methylpolyethylene glycol 5′000

Synonym: O-Methyl-O′-[2-(6-oxocaproylamino)ethyl]polyethylene glycol 5′000, PEG-aldehyde 5′000

Purchase

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

1 g in poly bottle

250 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?