• USA Home
  • 42020 - 2,4-Dinitrobenzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
42020 Sigma-Aldrich

2,4-Dinitrobenzenesulfonyl chloride

purum, ≥98.0% (AT)

  • CAS Number 1656-44-6

  • Linear Formula (O2N)2C6H3SO2Cl

  • Molecular Weight 266.62

  •  Beilstein Registry Number 2147583

  •  EC Number 216-749-3

  •  MDL number MFCD00007431

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
InChI Key   SSFSNKZUKDBPIT-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
mp   101-103 °C(lit.)
  99-103 °C

Description

Other Notes

Starting material for the preparation of 2,4-dinitro-benzenesulfonic acid hydrazide; "A useful reagent for the Eschenmoser, α,β-cleavage of α,β-epoxy ketones"

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

142956 2,4-Dinitrobenzenesulfonyl chloride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?