• USA Home
  • 42033 - N-(3,5-Dinitrobenzoyl)-L-α-phenylglycine

EMAIL THIS PAGE TO A FRIEND
42033 Sigma-Aldrich

N-(3,5-Dinitrobenzoyl)-L-α-phenylglycine

purum, ≥98.0% (sum of enantiomers, HPLC)

Synonym: (S)-(+)-N-(3,5-Dinitrobenzoyl)phenylglycine

  • CAS Number 90761-62-9

  • Empirical Formula (Hill Notation) C15H11N3O7

  • Molecular Weight 345.26

  •  Beilstein Registry Number 4719761

  •  MDL number MFCD00064470

  •  PubChem Substance ID 329756175

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chemical Biology, Chemical Synthesis, Derivatization Reagents,
InChI Key   MIVUDAUOXJDARR-ZDUSSCGKSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D +103±3°, c = 1% in THF
mp   215-217 °C (dec.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?