• USA Home
  • 42080 - 1-Fluoro-2,4-dinitrobenzene

EMAIL THIS PAGE TO A FRIEND
42080 Sigma-Aldrich

1-Fluoro-2,4-dinitrobenzene

≥99.0% (GC)

Synonym: 2,4-Dinitro-1-fluorobenzene, DNFB, DNPF, FDNB, Sanger reagent

  • CAS Number 70-34-8

  • Linear Formula (O2N)2C6H3F

  • Molecular Weight 186.10

  •  Beilstein Registry Number 398632

  •  EC Number 200-734-3

  •  MDL number MFCD00007056

  •  PubChem Substance ID 329756185

Purchase

Properties

Related Categories α-aminocarboxylic Acids, Amino Acid Analysis, Amino Acid Analysis Reagents, Amino Acids, Analytical Reagents,
InChI Key   LOTKRQAVGJMPNV-UHFFFAOYSA-N
assay   ≥99.0% (GC)
refractive index   n20/D 1.569(lit.)
  n20/D 1.570
bp   178 °C/25 mmHg(lit.)
mp   25-27 °C(lit.)
  25-27 °C
solubility   chloroform: soluble0.1 g/mL, clear
density   1.482 g/mL at 25 °C(lit.)

Description

Other Notes

For the modification and determination of amino acids, peptides and phenols ; As a probe for the presence of hydrodisulfide groups in proteins, For the detection of amphetamine enantiomers by circular dichroism spectroscopy.

This product has been replaced by D1529-SIGMA | 1-Fluoro-2,4-dinitrobenzene ≥99%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
327.2 °F
Flash Point(C) 
164 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?