• USA Home
  • 42110 - 1-Iodo-2,4-dinitrobenzene

EMAIL THIS PAGE TO A FRIEND
42110 Sigma-Aldrich

1-Iodo-2,4-dinitrobenzene

purum

  • CAS Number 709-49-9

  • Linear Formula IC6H3(NO2)2

  • Molecular Weight 294.00

  •  Beilstein Registry Number 1113584

  •  EC Number 211-908-3

  •  MDL number MFCD00039738

Purchase

Properties

InChI Key   FXMKXMJLXRTQSW-UHFFFAOYSA-N
grade   purum
mp   86-88 °C(lit.)
  86-88 °C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?