CAS Number 7681-52-9
Linear Formula NaClO
Molecular Weight 74.44
EC Number 231-668-3
MDL number MFCD00011120
PubChem Substance ID 24866608
Popular Documents: Specification Sheet (PDF)
| Related Categories | Chemical Synthesis, Essential Chemicals, Hypochlorites, Inorganic Salts, Solutions and Reagents, Oxidizing Agents, |
| grade | reagent grade |
| composition | available chlorine, 10-15% |
| bp | 111 °C |
| density | 1.206 g/mL at 25 °C |
| storage temp. | 2-8°C |
Live Chat and Frequently Asked Questions
are available for this Product.
1 L in poly bottle
250 mL in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?