• USA Home
  • 42554 - 1,2-Dipalmitoyl-rac-glycerol

EMAIL THIS PAGE TO A FRIEND
42554 Sigma-Aldrich

1,2-Dipalmitoyl-rac-glycerol

≥99.0% (after transesterification, GC)

Synonym: (±)-1,2-Dipalmitin, 1,2-Dihexadecanoyl-rac-glycerol, rac-Glycerol 1,2-dipalmitate, DL-1,2-Dipalmitin, DG(16:0/16:0/0:0)[rac], Hexadecanoic acid 1,1′-[1-(hydroxymethyl)-1,2-ethanediyl] ester

  • CAS Number 40290-32-2

  • Empirical Formula (Hill Notation) C35H68O5

  • Molecular Weight 568.91

  •  Beilstein Registry Number 1809777

  •  EC Number 254-870-3

  •  MDL number MFCD00042874

  •  PubChem Substance ID 329756237

Purchase

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?