EMAIL THIS PAGE TO A FRIEND
42560 Sigma-Aldrich

Dipentene

technical, for use as solvent (for the paint industry), mixture of various terpenes

Synonym: (±)-Limonene, p-Mentha-1,8-diene

  • CAS Number 138-86-3

  • Empirical Formula (Hill Notation) C10H16

  • Molecular Weight 136.23

  •  Beilstein Registry Number 3587825

  •  EC Number 205-341-0

  •  MDL number MFCD00062992

  •  PubChem Substance ID 329756238

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
grade   for use as solvent (for the paint industry)
vapor density   >4.7 (80 °C, vs air)
vapor pressure   1 mmHg ( 20 °C)
InChI Key   XMGQYMWWDOXHJM-UHFFFAOYSA-N
grade   technical
autoignition temp.   458 °F
expl. lim.   0.7-6.1 %, 150 °F
refractive index   n20/D 1.473(lit.)
  n20/D 1.473
bp   170-180 °C(lit.)
density   0.86 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2052 3 / PGIII
WGK Germany 
3
RTECS 
OS8100000
Flash Point(F) 
109.4 °F
Flash Point(C) 
43 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

334111 Dipentene, technical, for use as solvent (for the paint industry), mixture of various terpenes

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?