• USA Home
  • 42815 - Diphenylborinic anhydride

EMAIL THIS PAGE TO A FRIEND
42815 Sigma-Aldrich

Diphenylborinic anhydride

purum p.a., for fluorescence, ≥95.0% (T)

Synonym: Oxybis(diphenylborane), Tetraphenyldiboroxane

  • CAS Number 4426-21-5

  • Linear Formula (C6H5)2BOB(C6H5)2

  • Molecular Weight 346.04

  •  Beilstein Registry Number 2950331

  •  MDL number MFCD00042663

  •  PubChem Substance ID 24866798

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Carbohydrate Labels, Carbohydrate Labels for Chromatography, Detection,
grade   for fluorescence
InChI Key   SNQFEECGHGUHBK-UHFFFAOYSA-N
grade   purum p.a.
assay   ≥95.0% (T)
mp   135-140 °C(lit.)
  135-140 °C
storage temp.   2-8°C

Description

Other Notes

Reagent used in the fluorometric determination of primary amines by thin-layer chromatography ; and high-performance liquid chromatography

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?