EMAIL THIS PAGE TO A FRIEND
42880 Sigma-Aldrich

Diphenylmethanol

puriss., ≥99.0% (GC)

Synonym: Benzhydrol, Benzhydryl alcohol, Diphenyl carbinol

  • CAS Number 91-01-0

  • Linear Formula (C6H5)2CHOH

  • Molecular Weight 184.23

  •  Beilstein Registry Number 1424379

  •  EC Number 202-033-8

  •  MDL number MFCD00004488

  •  PubChem Substance ID 329756276

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, Chemical Synthesis, Organic Building Blocks,
InChI Key   QILSFLSDHQAZET-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
bp   297-298 °C(lit.)
mp   65-67 °C(lit.)
  65-67 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DC7452000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?