• USA Home
  • 428914 - Palladium matrix modifier solution

EMAIL THIS PAGE TO A FRIEND
428914 Sigma-Aldrich

Palladium matrix modifier solution

2% Pd in 1-3 wt.% HNO3, 99.999%

  •  eCl@ss 38191001

Purchase

Description

Application

For graphite furnace analyses.

Features and Benefits

Chloride free.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Optimizing Analysis Conditions in AAS Measurements

As your one-stop provider for instrumental analysis via Atomic Absorption Spectroscopy (AAS), we offer a comprehensive choice of reagents for use in sample preparation as well as in calibration. Our ...
Keywords: Absorption, Atomic absorption spectroscopy, Buffers, Phase transitions, Sample preparations, Spectroscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?