• USA Home
  • 42910 - Diphenylcyclopropenone

EMAIL THIS PAGE TO A FRIEND
42910 Sigma-Aldrich

Diphenylcyclopropenone

purum, ≥98.0% (HPLC)

Synonym: DPC

  • CAS Number 886-38-4

  • Empirical Formula (Hill Notation) C15H10O

  • Molecular Weight 206.24

  •  Beilstein Registry Number 608049

  •  EC Number 212-948-4

  •  MDL number MFCD00001311

  •  PubChem Substance ID 329756281

Purchase

Properties

Related Categories Building Blocks, C15 to C38, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   HCIBTBXNLVOFER-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   118-122 °C(lit.)
  118-122 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

177377 Diphenylcyclopropenone, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?