EMAIL THIS PAGE TO A FRIEND
42960 Sigma-Aldrich

Diphenyl disulfide

technical, ≥97.0% (HPLC)

Synonym: Phenyl disulfide

  • CAS Number 882-33-7

  • Linear Formula C6H5SSC6H5

  • Molecular Weight 218.34

  •  Beilstein Registry Number 639794

  •  EC Number 212-926-4

  •  MDL number MFCD00003065

  •  PubChem Substance ID 24866872

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfides/Disulfides, Sulfur Compounds More...
InChI Key   GUUVPOWQJOLRAS-UHFFFAOYSA-N
grade   technical
assay   ≥97.0% (HPLC)
mp   58-60 °C

Description

Other Notes

Reagent for the α-phenylsulfenylation of carbonyl compounds

This product has been replaced by 169021-ALDRICH | Phenyl disulfide 99%

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN3077 - class 9 - PG 3 - DOT/IATA UN3335 - Environmentally hazardous substances, solid, n.o.s., HI: all (not BR)
WGK Germany 
3
RTECS 
SS6825000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

169021 Phenyl disulfide, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?