EMAIL THIS PAGE TO A FRIEND
43181 Sigma-Aldrich

2,2-Diphenylpropane

purum, ≥96.0% (GC)

  • CAS Number 778-22-3

  • Linear Formula (CH3)2C(C6H5)2

  • Molecular Weight 196.29

  •  Beilstein Registry Number 2041894

  •  EC Number 212-292-9

  •  MDL number MFCD00008794

  •  PubChem Substance ID 24867048

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   MILSYCKGLDDVLM-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
refractive index   n20/D 1.57(lit.)
bp   282 °C(lit.)
mp   26-28 °C(lit.)
  26-28 °C
density   0.992 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?