• USA Home
  • 43565 - Dipropylene glycol monomethyl ether

EMAIL THIS PAGE TO A FRIEND
43565 Sigma-Aldrich

Dipropylene glycol monomethyl ether

purum, mixture of isomers, ≥95.0% (GC)

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
InChI Key   QCAHUFWKIQLBNB-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.422
bp   90-91 °C/12 mmHg(lit.)
density   0.954 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NA 1993 / PGIII
WGK Germany 
3
RTECS 
JM1575000
Flash Point(F) 
165.2 °F
Flash Point(C) 
74 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

283282 Di(propylene glycol) methyl ether, mixture of isomers, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?