437565 Sigma-Aldrich

1,2-Dichloroethane

ACS reagent, ≥99%

DOWNLOAD MSDS (PDF)

Synonym: Ethylene chloride Ethylene dichloride

  • CAS Number 107-06-2

  • Linear Formula ClCH2CH2Cl

  • Molecular Weight 98.96

  •  Beilstein Registry Number 605264

  •  EC Number 203-458-1

  •  MDL number MFCD00000963

Properties

grade   ACS reagent
vapor density   3.4 (20 °C, vs air)
vapor pressure   87 mmHg ( 25 °C)
assay   ≥99%
autoignition temp.   775 °F
expl. lim.   15.6 %
total impurities   ≤0.0003 meq/g Titr. Acid
  ≤0.03% water
evapn. residue   ≤0.002%
color   APHA: ≤10
refractive index   n20/D 1.444(lit.)
bp   83 °C(lit.)
  83 °C
mp   −35 °C(lit.)
density   1.256 g/mL at 25 °C(lit.)

Price and Availability

Customers Also Viewed

anhydrous, 99.8%

ACS reagent, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?