EMAIL THIS PAGE TO A FRIEND
437603 Sigma-Aldrich

1-Butanol

ACS reagent, ≥99.4%

Synonym: n-Butanol, Butyl alcohol

  • CAS Number 71-36-3

  • Linear Formula CH3(CH2)3OH

  • Molecular Weight 74.12

  •  Beilstein Registry Number 969148

  •  EC Number 200-751-6

  •  MDL number MFCD00002964

  •  PubChem Substance ID 329756390

Purchase

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, PVC Coated Bottles, Solvent Bottles,
grade   ACS reagent
vapor density   2.55 (vs air)
vapor pressure   5.5 mmHg ( 20 °C)
InChI Key   LRHPLDYGYMQRHN-UHFFFAOYSA-N
assay   ≥99.4%
autoignition temp.   649 °F
expl. lim.   11.2 %
impurities   ≤0.0008 meq/g Titr. Acid
  ≤0.01% butyraldehyde
  ≤0.1% water
  ≤0.2% butyl ether
evapn. residue   ≤0.005%
color   APHA: ≤10
refractive index   n20/D 1.399(lit.)
bp   116-118 °C(lit.)
  117.7 °C
mp   −90 °C(lit.)
density   0.81 g/mL at 25 °C(lit.)

Description

Packaging

Packaged in poly-coated bottles

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1120 - class 3 - PG 3 - Butanols
WGK Germany 
1
RTECS 
EO1400000
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?