• USA Home
  • 43789 - 3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester)

EMAIL THIS PAGE TO A FRIEND
43789 Sigma-Aldrich

3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester)

≥97.0% (HPLC)

Synonym: DTSP, Di(N-succinimidyl) 3,3′-dithiodipropionate, Dithiobis(succinimidyl propionate), Lomant’s reagent

  • CAS Number 57757-57-0

  • Empirical Formula (Hill Notation) C14H16N2O8S2

  • Molecular Weight 404.42

  •  Beilstein Registry Number 1518074

  •  EC Number 260-931-5

  •  MDL number MFCD00042045

  •  PubChem Substance ID 329756399

Purchase

Properties

InChI Key   FXYPGCIGRDZWNR-UHFFFAOYSA-N
assay   ≥97.0% (HPLC)
mp   128-133 °C
storage temp.   2-8°C

Description

Other Notes

Cleavable cross-linking reagent

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D3669 3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?