EMAIL THIS PAGE TO A FRIEND
43800 Sigma-Aldrich

Dithiooxamide

for spectrophotometric det. of Cu, Os, Bi, Co, Cu, Fe, Ni, Os, Pt, Ru, ≥98.5%

Synonym: Dithiooxalic diamide, Rubeanic acid

  • CAS Number 79-40-3

  • Linear Formula NH2CSCSNH2

  • Molecular Weight 120.20

  •  Beilstein Registry Number 605577

  •  EC Number 201-203-9

  •  MDL number MFCD00004941

  •  PubChem Substance ID 329756404

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, C-D, Puriss p.a., Puriss p.a.,
InChI Key   OAEGRYMCJYIXQT-UHFFFAOYSA-N
assay   ≥98.5% (N)
  ≥98.5%
quality   for spectrophotometric det. of Cu, Os, Bi, Co, Cu, Fe, Ni, Os, Pt, Ru
ign. residue   ≤0.1%
mp   ≥300 °C(lit.)

Description

General description

Dithiooxamide is an organic compound which is basically a sulfur analog of oxamide. It is mostly used as metal deactivators and vulcanization accelerators. It forms stable metal complexes which acts as colour sources in duplicating processes.

Price and Availability


Laboratory Gloves

Spectroquant Prove
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RP1575000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?