CAS Number 7697-37-2
Empirical Formula (Hill Notation) HNO3
Molecular Weight 63.01
EC Number 231-714-2
MDL number MFCD00011349
PubChem Substance ID 24867421
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, |
| grade | ACS reagent |
| vapor pressure | 8 mmHg ( 20 °C) |
| assay | 68.0-70.0% (ACS specification) |
| 70% | |
| ign. residue | ≤5 ppm |
| color | APHA: ≤10 |
| bp | 120.5 °C(lit.) |
2.5 L in glass bottle
500 mL in glass bottle
6×2.5 L in PVC coated
6×500 mL in PVC coated
Eliminate repetitive Molar calculations and potential errors with these conveniently designed graduated cylinders. They featureCustomers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?