• USA Home
  • 43821 - (−)-O,O′-Di-p-toluoyl-L-tartaric acid

EMAIL THIS PAGE TO A FRIEND
43821 Sigma-Aldrich

(−)-O,O′-Di-p-toluoyl-L-tartaric acid

purum, ≥98.0% (T)

  • CAS Number 32634-66-5

  • Empirical Formula (Hill Notation) C20H18O8

  • Molecular Weight 386.35

  •  Beilstein Registry Number 2022481

  •  EC Number 251-131-7

  •  MDL number MFCD00064440

  •  PubChem Substance ID 24867435

  •  eCl@ss 39021705

Purchase

Properties

Related Categories Asymmetric Synthesis, Carboxylic Acids, Chemical Synthesis, Chiral Building Blocks, Chiral Resolution Reagents,
InChI Key   CMIBUZBMZCBCAT-HZPDHXFCSA-N
grade   purum
assay   ≥98.0% (T)
optical activity   [α]20/D −139±2°, c = 1% in ethanol

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?