Synonym: EtOAc
CAS Number 141-78-6
Linear Formula CH3COOC2H5
Molecular Weight 88.11
Beilstein Registry Number 506104
EC Number 205-500-4
MDL number MFCD00009171
Popular Documents: Specification Sheet (PDF)
| Related Categories | Analytical Reagents, Analytical/Chromatography, CHROMASOLV for HPLC, Chromatography Reagents & Solvents, Composite Drums, |
| grade | CHROMASOLV® |
| for HPLC | |
| vapor density | 3 (20 °C, vs air) |
| vapor pressure | 73 mmHg ( 20 °C) |
| assay | ≥99.8% |
| autoignition temp. | 801 °F |
| expl. lim. | 2.2-11.5 %, 38 °F |
2×10, 20 L in Nowpak™
4, 4×4 L in PVC coated
50 L in comp drum
CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?