EMAIL THIS PAGE TO A FRIEND
43920 Sigma-Aldrich

Divinyl sulfone

purum, ≥98.0% (GC)

Synonym: Vinyl sulfone

  • CAS Number 77-77-0

  • Linear Formula (CH2=CH)2SO2

  • Molecular Weight 118.15

  •  Beilstein Registry Number 1071329

  •  EC Number 201-057-6

  •  MDL number MFCD00008623

  •  PubChem Substance ID 329756427

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfones, Sulfur Compounds More...
InChI Key   AFOSIXZFDONLBT-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
contains   ~0.05% hydroquinone as stabilizer
refractive index   n20/D 1.476(lit.)
  n20/D 1.477
bp   234 °C(lit.)
mp   −26 °C(lit.)
density   1.177 g/mL at 25 °C(lit.)
storage temp.   2-8°C
Gene Information   human ... LOC129293(129293)

Description

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2810 6.1/PG 1
WGK Germany 
3
RTECS 
KM7175000
Flash Point(F) 
216 °F
Flash Point(C) 
102 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

V3700 Divinyl sulfone, contains <650 ppm hydroquinone as inhibitor, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?