EMAIL THIS PAGE TO A FRIEND
43960 Sigma-Aldrich

1-Docosanol

purum, ≥97.0% (GC)

Synonym: Behenyl alcohol

  • CAS Number 661-19-8

  • Linear Formula CH3(CH2)21OH

  • Molecular Weight 326.60

  •  Beilstein Registry Number 1770470

  •  EC Number 211-546-6

  •  MDL number MFCD00002939

  •  PubChem Substance ID 329756435

Purchase

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C11 to C30+, Cell Biology,
InChI Key   NOPFSRXAKWQILS-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   180 °C/0.22 mmHg(lit.)
mp   65-72 °C(lit.)
  70-71 °C

Price and Availability


The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

169102 1-Docosanol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?