EMAIL THIS PAGE TO A FRIEND
44130 Sigma-Aldrich

1-Dodecanethiol

purum, ≥97.0% (GC)

Synonym: n-Dodecyl mercaptan, Dodecyl mercaptan, Lauryl mercaptan, Mercaptan C12, NDM

  • CAS Number 112-55-0

  • Linear Formula CH3(CH2)11SH

  • Molecular Weight 202.40

  •  Beilstein Registry Number 969337

  •  EC Number 203-984-1

  •  MDL number MFCD00004885

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
vapor density   7 (vs air)
InChI Key   WNAHIZMDSQCWRP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.459(lit.)
  n20/D 1.459
bp   266-283 °C(lit.)
density   0.845 g/mL at 25 °C(lit.)

Description

Other Notes

For the regeneration of native proteins from mercuribenzoate

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1760 - class 8 - PG 3 - acidic - EHS - Corrosive liquids, n.o.s., HI: all
WGK Germany 
3
RTECS 
JR3155000
Flash Point(F) 
248 °F
Flash Point(C) 
120 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

471364 1-Dodecanethiol, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?