EMAIL THIS PAGE TO A FRIEND
44150 Sigma-Aldrich

1-Dodecene

technical, ≥90% (GC)

  • CAS Number 112-41-4

  • Linear Formula CH3(CH2)9CH=CH2

  • Molecular Weight 168.32

  •  Beilstein Registry Number 1699848

  •  EC Number 203-968-4

  •  MDL number MFCD00008961

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
vapor density   5.8 (vs air)
vapor pressure   0.2 mmHg ( 20 °C)
InChI Key   CRSBERNSMYQZNG-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
autoignition temp.   491 °F
expl. lim.   ~4.7 %
refractive index   n20/D 1.429(lit.)
  n20/D 1.430
bp   214-216 °C(lit.)
mp   −35 °C(lit.)
density   0.758 g/mL at 20 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
WGK Germany 
3
Flash Point(F) 
163.4 °F
Flash Point(C) 
73 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?