443778 Sigma-Aldrich

1-Methyl-2-pyrrolidinone

ACS reagent, ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: 1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

Properties

Related Categories ACS Grade, ACS Grade Solvents, ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Cans with NPT Threads,
grade   ACS reagent
vapor density   3.4 (vs air)
vapor pressure   0.29 mmHg ( 20 °C)
  0.99 mmHg ( 40 °C)
assay   ≥99.0%
autoignition temp.   518 °F
expl. lim.   9.5 %
impurities   ≤0.01% Free amines (as CH3NH2)
  ≤0.05% water
color   APHA: ≤50
refractive index   n20/D 1.47(lit.)
bp   202 °C(lit.)
  81-82 °C/10 mmHg(lit.)
mp   −24 °C(lit.)
density   1.028 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤1 ppm

Description

Packaging

1, 2.5 L in glass bottle

View returnable container options.

20 L in steel drum

500 mL in glass bottle

Price and Availability

Customers Also Viewed

1-<WBR>Methyl-<WBR>2-<WBR>pyrrolidinone

CHROMASOLV® Plus, for HPLC, ≥99%

1-<WBR>Methyl-<WBR>2-<WBR>pyrrolidinone

anhydrous, 99.5%

1-<WBR>Methyl-<WBR>2-<WBR>pyrrolidinone

biotech. grade, ≥99.5%

1-<WBR>Methyl-<WBR>2-<WBR>pyrrolidinone

spectrophotometric grade, ≥99%

1-<WBR>Methyl-<WBR>2-<WBR>pyrrolidinone

puriss. p.a., ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?