EMAIL THIS PAGE TO A FRIEND
44380 Sigma-Aldrich
Dowex® 2X8
Cl--form, strongly basic
Purchase
Properties
Related Categories Analytical/Chromatography, Anion Exchange Media, Dowex Strong Anion Exchangers, Resins & LC Separation Media, Resins and Media More...
loss   ~40% loss on drying, 110 °C
particle size   50-100 mesh
Description
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Legal Information
DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Articles
Selection Guide for Polymeric Resins in Chromatography & Purification
Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?