EMAIL THIS PAGE TO A FRIEND
44435 Sigma-Aldrich

Dowex® 21K

Cl--form, strongly basic

Purchase

Properties

loss   ~55% loss on drying, 110 °C
particle size   20-50 mesh

Description

General description

also called SBR-P

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Legal Information

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

436658 Dowex® 21K chloride form, chloride form, 16-30 mesh

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?