EMAIL THIS PAGE TO A FRIEND
44443 Sigma-Aldrich
Dowex® 50X8
H+-form, strongly acidic
Purchase
Properties
loss   ~50% loss on drying, 110 °C
particle size   20-50 mesh
Description
General description
also called HCR-S
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Legal Information
DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Price and Availability
Safety & Documentation
Safety Information
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
433950 Dowex® Marathon C hydrogen form, 23-27 mesh particle size
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?