EMAIL THIS PAGE TO A FRIEND
44460 Sigma-Aldrich

Dowex® 50WX2

H+-form, strongly acidic

Purchase

Properties

Related Categories Analytical/Chromatography, Cation Exchange Media, Dowex Strong Cation Exchangers, Resins & LC Separation Media, Resins and Media More...
cross-linkage   2%
moisture   74-82%
loss   ~80% loss on drying, 110 °C
matrix   styrene divinylbenzene (gel)
particle size   100-200 mesh
capacity   0.6 meq/mL by wetted bed volume

Description

Legal Information

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Selection Guide for Polymeric Resins in Chromatography & Purification

Sigma-Aldrich carries a vast array of resins, from leading manufacturers; such as, Dow Chemical, Rohm & Haas, Mitsubishi, and Lanxess. Our product line includes adsorbent, ion-exchange (anion / catio...
Keywords: Adsorption, Antibiotics, Detergents, Fermentation, Filtration, Hormones, Ion Exchange, Pharmaceutical, Purification, Separation

Related Products

replaced by

Product #

Description

Add to Cart

217468 Dowex® 50WX2 hydrogen form, 100-200 mesh

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?