• USA Home
  • 44552 - Dowex® DR-2030 hydrogen form

EMAIL THIS PAGE TO A FRIEND
44552 Sigma-Aldrich

Dowex® DR-2030 hydrogen form

H+-form, strongly acidic

Purchase

Properties

quality   dry
parameter   150 °C max. temp.
moisture   ~3%
loss   ≤3% loss on drying, 110 °C
matrix   styrene-divinylbenzene (macroporous)
matrix active group   sulfonic acid functional group
particle size   20-50 mesh
operating pH   0 - 14
capacity   1.7 meq/mL by wetted bed volume
  4.7 meq/g by dry weight

Description

Components

Made from Dowex M-31.

Application

Strong acid catalyst. Does NOT meet FDA Food Additive Regulation 21 CFR 173.25.

suitable for nonaqueous systems

Legal Information

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?