EMAIL THIS PAGE TO A FRIEND
44557 Sigma-Aldrich

Dowex® MSC-1

strongly acidic, Na+-form, macroporous

Purchase

Properties

form   macroporous
loss   ~50% loss on drying, 110 °C
particle size   20-50 mesh

Description

Legal Information

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?