EMAIL THIS PAGE TO A FRIEND
44656 Sigma-Aldrich

α-Ecdysone

≥99.0% (UV)

Synonym: 2β,3β,14α,22(R),25-Pentahydroxy-7-cholesten-6-one

  • CAS Number 3604-87-3

  • Empirical Formula (Hill Notation) C27H44O6

  • Molecular Weight 464.63

  •  Beilstein Registry Number 2422986

  •  EC Number 222-760-4

  •  MDL number MFCD00042683

  •  PubChem Substance ID 329756531

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Growth regulators, Insecticides, Pesticides & Metabolites Standards More...
InChI Key   UPEZCKBFRMILAV-JMZLNJERSA-N
assay   ≥99.0% (UV)
storage temp.   −20°C

Description

General description

Prothoracic hormone (causes moulting in insects)

Other Notes

Prothorakal gland hormone

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
FZ8170000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

E9004 α-Ecdysone, ≥90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?