EMAIL THIS PAGE TO A FRIEND
44657 Sigma-Aldrich

ECD MUG Agar

for microbiology

Purchase

Properties

Related Categories A - M, All Food Control Media (alphabetical), All Solid Media/Agars, Escherichia coli & Coliforms, Fluorogenic Media,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 15 g/L
  bile salt mixture, 1.5 g/L
  casein peptone, 20 g/L
  dipotassium hydrogen phosphate, 4 g/L
  lactose, 5 g/L
  4-methylumbelliferyl-β-D-glucuronide, 0.07 g/L
  potassium hydrogen phosphate, 1.5 g/L
  sodium chloride, 5 g/L
  tryptophan, 1 g/L
final pH   7.0±0.2 (25 °C)
suitability   selective and differential for Escherichia coli and Coliforms

Description

Application

The bile-salt mixture in this E. coli Direct Agar extensively inhibits the non-obligatory intestinal accompanying flora. Fluorescence in the UV and a positive indole test demonstrate the presence of E. coli in the colonies.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?