• USA Home
  • 4466096 - iTraq® Reagent 25U Kit

EMAIL THIS PAGE TO A FRIEND
4466096 Sigma-Aldrich

iTraq® Reagent 25U Kit

Purchase

Properties

Related Categories Mass Spectrometry, Molecular Biology, Proteomics, Sciex iChemistry Solutions, iTRAQ Reagents More...
shipped in   dry ice
storage temp.   −20°C

Description

General description

Includes 4 tubes. Each tube contains 25 units of an individual 4-plex reagent (iTRAQ Reagents 114, 115, 116, 117).

Legal Information

iTraq is a registered trademark of AB Sciex Pte. Ltd.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?