EMAIL THIS PAGE TO A FRIEND
44660 Sigma-Aldrich

Fast Blue B Salt

for microscopy (Hist.), for TLC-detection of aflatoxins

Synonym: o-Dianisidine bis(diazotized) zinc double salt, Azoic Diazo No. 48, DBB, Diazo Blue B, Naphthanil Diazo Blue B

  • CAS Number 14263-94-6

  • Linear Formula C14H12N4O2Cl2 · ZnCl2

  • Molecular Weight 475.47

  •  Beilstein Registry Number 1499067

  •  Colour Index Number 37235

  •  EC Number 238-153-2

  •  MDL number MFCD00075203

Purchase

Properties

grade   for microscopy (Hist.)
InChI Key   GPPKNJIWDULNQH-UHFFFAOYSA-J
quality   for TLC-detection of aflatoxins
mp   >300 °C(lit.)

Description

Other Notes

Stain used for the determination of various compounds, e.g. proteins; bilirubin; enzymes;

This product has been replaced by D9805-Sigma-Aldrich | Fast Blue B Salt Dye content ~95 %

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D9805 Fast Blue B Salt, Dye content ~95 %

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?