EMAIL THIS PAGE TO A FRIEND
44750 Sigma-Aldrich

Fast Red RC Salt

for microscopy (Hist.)

Synonym: 2-Amino-4-chloroanisole diazotated zinc chloride complex, 5-Chloro-2-methoxybenzenediazonium chloride hemi(zinc chloride) salt, Diazo Fast Red RC

  • CAS Number 68025-25-2

  • Empirical Formula (Hill Notation) C7H6Cl2N2O · 0.5ZnCl2

  • Molecular Weight 273.19

  •  Colour Index Number 37120

  •  EC Number 268-183-1

  •  MDL number MFCD00074764

Purchase

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?