EMAIL THIS PAGE TO A FRIEND
44760 Sigma-Aldrich

Fast Black K salt

for microscopy

Synonym: 2,5-Dimethoxy-4-(4-nitrophenylazo)benzenediazonium chloride zinc double salt, 4-Amino-2,5-dimethoxy-4′-nitroazobenzene diazotated zinc double salt, Azoic Diazo No. 38

  • CAS Number 64071-86-9

  • Empirical Formula (Hill Notation) C14H12ClN5O4 · 0.5ZnCl2

  • Molecular Weight 417.88

  •  Colour Index Number 37190

  •  EC Number 264-656-1

  •  MDL number MFCD00078471

  •  PubChem Substance ID 329756542

Purchase

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

F7253 Fast Black K Salt hemi(zinc chloride) salt, practical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?