EMAIL THIS PAGE TO A FRIEND
44860 Sigma-Aldrich

1-Eicosanol

purum, ≥97.0% (GC)

Synonym: Arachinyl alcohol

  • CAS Number 629-96-9

  • Linear Formula CH3(CH2)19OH

  • Molecular Weight 298.55

  •  Beilstein Registry Number 1705104

  •  EC Number 211-119-4

  •  MDL number MFCD00002938

  •  PubChem Substance ID 329756553

Purchase

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C11 to C30+, Cell Biology,
vapor density   7.4 (vs air)
vapor pressure   <0.1 mmHg ( 20 °C)
InChI Key   BTFJIXJJCSYFAL-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
mp   62-65 °C(lit.)
  63-66 °C
solubility   water: soluble0.001 g/L at 23 °C

Price and Availability


Laboratory Gloves

The Future of Bioimaging

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
381.2 °F
Flash Point(C) 
194 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?