• USA Home
  • 44982 - Iron(II) sulfate heptahydrate

EMAIL THIS PAGE TO A FRIEND
44982 Sigma-Aldrich

Iron(II) sulfate heptahydrate

tested according to Ph.Eur.

Synonym: Ferrosi sulfas, Ferrous sulfate heptahydrate

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z More...
vapor pressure   14.6 mmHg ( 25 °C)
InChI Key   SURQXAFEQWPFPV-UHFFFAOYSA-L
pharmacopeia   testing conforms to Pharmacopeia
density   1.898 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
NO8510000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?