• USA Home
  • 45175 - Endoproteinase Lys-C from Lysobacter enzymogenes

EMAIL THIS PAGE TO A FRIEND
45175 Sigma-Aldrich

Endoproteinase Lys-C from Lysobacter enzymogenes

~3 U/vial

Purchase

Description

Other Notes

Lysine specific protease, used in protein sequence analysis; Site-directed limited proteolysis

Same product — same quality — now Sigma-Aldrich brand

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol 4-nitroaniline per minute at 25 °C (Chromozym® PL as substrate, ® registered trademark of Pentapharm AG, Basel).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

P2289 Endoproteinase Lys-C from Lysobacter enzymogenes, lyophilized powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?