EMAIL THIS PAGE TO A FRIEND
45187 Sigma-Aldrich

Endothelin 1

≥97.0% (HPLC)

Synonym: ET-1

  • CAS Number 117399-94-7

  • Empirical Formula (Hill Notation) C109H159N25O32S5

  • Molecular Weight 2491.90

  •  MDL number MFCD00133305

  •  PubChem Substance ID 24868658

Purchase

Properties

Related Categories Agonists, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
InChI Key   ZUBDGKVDJUIMQQ-ZTNLKOGPSA-N
assay   ≥97.0% (HPLC)
storage temp.   −20°C
Gene Information   human ... EDN1(1906)

Description

Amino Acid Sequence

Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp [Disulfide Bridges: 1-15; 3-11]

Other Notes

A synthetic peptide with the sequence of human and porcine Endothelin 1.

Review. Endothelins as regulators of vascular tone in man

Same product — same quality — now Sigma-Aldrich brand

Biochem/physiol Actions

Potent agonist for ETA and ETB endothelin receptors. Induces the production of hypoxia-inducible factor 1α, and, thus, the production of VEGF.

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KE9446500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

E7764 Endothelin 1, ≥97% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?