EMAIL THIS PAGE TO A FRIEND
45188 Sigma-Aldrich

Endothelin 2, human

≥97.0% (HPLC)

Purchase

Properties

Related Categories Agonists, Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes,
InChI Key   MLFJHYIHIKEBTQ-IYRKOGFYSA-N
assay   ≥97.0% (HPLC)
storage temp.   −20°C
Gene Information   human ... EDN2(1907)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGI
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?