• USA Home
  • 45280 - (+)-Ephedrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
45280 Sigma-Aldrich

(+)-Ephedrine hydrochloride

purum, ≥99.0% (AT)

Synonym: (1S,2R)-2-Methylamino-1-phenyl-1-propanol hydrochloride, D-α-(1-Methylaminoethyl)benzyl alcohol hydrochloride

  • CAS Number 24221-86-1

  • Empirical Formula (Hill Notation) C10H15NO · HCl

  • Molecular Weight 201.69

  •  Beilstein Registry Number 6118541

  •  EC Number 246-090-7

  •  MDL number MFCD00064551

  •  PubChem Substance ID 329756627

Purchase

Properties

Related Categories Alkaloid, Asymmetric Synthesis, Cell Biology, Chemical Synthesis, Chiral Auxiliaries,
InChI Key   BALXUFOVQVENIU-GHXDPTCOSA-N
grade   purum
assay   ≥99.0% (AT)
optical activity   [α]20/D +34±1°, c = 11.5% in H2O
mp   217-221 °C

Description

Other Notes

Sales restrictions may apply

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?