EMAIL THIS PAGE TO A FRIEND
45326 Sigma-Aldrich

Aniline

PESTANAL®, analytical standard

Purchase

Properties

Related Categories A-B, Additional Standards, Alpha Index of All Pesticide Standards, Amines, Analytical Standards,
grade   PESTANAL®, analytical standard
vapor density   3.22 (185 °C, vs air)
vapor pressure   0.7 mmHg ( 25 °C)
InChI Key   PAYRUJLWNCNPSJ-UHFFFAOYSA-N
form   neat
autoignition temp.   1139 °F
expl. lim.   11 %
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.586(lit.)
bp   184 °C(lit.)
mp   −6 °C(lit.)
density   1.022 g/mL at 25 °C(lit.)
format   neat

Description

Biochem/physiol Actions

The acute toxicity of aniline involves its activation in vivo to 4-hydroxyaniline and the formation of adducts with hemoglobin. In erythrocytes, this is associated with the release of iron and the accumulation of methemoglobin and the development of hemolytic anemia and inflammation of the spleen. Tumor formation is often observed in the spleen on prolonged administration.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
Target organs 
Blood
RIDADR 
UN 1547 6.1 / PGII
WGK Germany 
3
RTECS 
BW6650000
Flash Point(F) 
158 °F
Flash Point(C) 
70 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?