EMAIL THIS PAGE TO A FRIEND
45358 Sigma-Aldrich

Bromopyrazone

PESTANAL®, analytical standard

Synonym: 5-Amino-4-bromo-2-phenyl-(2H)-pyridazin-3-one

  • CAS Number 3042-84-0

  • Empirical Formula (Hill Notation) C10H8BrN3O

  • Molecular Weight 266.09

  •  Beilstein Registry Number 612440

  •  EC Number 221-248-8

  •  MDL number MFCD00055401

  •  PubChem Substance ID 329756672

Purchase

Properties

Related Categories A-B, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Herbicides,
grade   PESTANAL®, analytical standard
InChI Key   ODNZLRLWXRXPOH-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?