EMAIL THIS PAGE TO A FRIEND
45390 Sigma-Aldrich

Chlorophacinone

PESTANAL®, analytical standard

  • CAS Number 3691-35-8

  • Empirical Formula (Hill Notation) C23H15ClO3

  • Molecular Weight 374.82

  •  Beilstein Registry Number 2063081

  •  EC Number 223-003-0

  •  MDL number MFCD00055360

  •  PubChem Substance ID 24868865

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, C, Food and Cosmetic Component Standards,
grade   analytical standard
InChI Key   UDHXJZHVNHGCEC-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Other Notes

Same product — same quality — now Sigma brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 1 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
NK5335000
Flash Point(F) 
>212 °F
Flash Point(C) 
>100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?